missing translation for 'onlineSavingsMsg'
Learn More

Tarloxotinib bromide, MedChemExpress

Product Code. 16453586
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16453586 25 mg 25mg
16487406 100 mg 100mg
16483457 50 mg 50mg
16437438 10 mg 10mg
16419917 5 mg 5mg
5 options
This item is not returnable. View return policy

Product Code. 16453586

Brand: Medchem Express HY17632

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tarloxotinib bromide (TH-4000) is an irreversible EGFR/HER2 inhibitor.

Tarloxotinib bromide (TH-4000) is an irreversible EGFR/HER2 inhibitor.

Specifications

Chemical Name or Material Tarloxotinib bromide
CAS 1636180-98-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H24Br2ClN9O3
Quantity 25 mg
Synonym TH-4000
Solubility Information DMSO : ≥ 33 mg/mL (48.40 mM)
SMILES BrC1=C(Cl)C=CC(NC2=NC=NC3=C2C=C(NC(/C=C/C[N+](C)(C)CC4=C([N+]([O-])=O)N=CN4C)=O)N=C3)=C1.[Br-]
Molecular Weight (g/mol) 681.77
Formula Weight 681.77
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.