missing translation for 'onlineSavingsMsg'
Learn More

(-)-Talarozole, MedChemExpress

Product Code. 16413446
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16413446 25 mg 25mg
16481439 1 mg 1mg
16468777 5 mg 5mg
16466178 10 mg 10mg
16432317 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16413446 Supplier Medchem Express Supplier No. HY14802D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (-)-Talarozole is a potent inhibitor of retinoic acid metabolism extracted from patent WO 1997049704 A1.

(-)-Talarozole is a potent inhibitor of retinoic acid metabolism extracted from patent WO 1997049704 A1.

Specifications

Chemical Name or Material (-)-Talarozole
Physical Form Solid
CAS 201410-67-5
Color Off-White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C21H23N5S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 72.5 mg/mL (192.05 mM)
SMILES CCC(CC)C(C1=CC=C(NC2=NC3=CC=CC=C3S2)C=C1)N4N=CN=C4.[(-)].[Rotation]
Molecular Weight (g/mol) 377.51
Formula Weight 377.51
Percent Purity 98.02%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.