missing translation for 'onlineSavingsMsg'
Learn More

SBC-110736, MedChemExpress

Product Code. 16412786
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16412786

Brand: Medchem Express HY101832

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SBC-110736 is a proprotein convertase subtilisin kexin type 9 (PCSK9) inhibitor extracted from patent WO2014150395A1, Figure 1.

SBC-110736 is a proprotein convertase subtilisin kexin type 9 (PCSK9) inhibitor extracted from patent WO2014150395A1, Figure 1.

Specifications

Chemical Name or Material SBC-110736
Physical Form Solid
CAS 1629166-02-4
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C26H27N3O2
For Use With (Application) Metabolism-protein/nucleotide metabolism
Solubility Information DMSO : ≥ 100 mg/mL (241.83 mM)
SMILES CC(NC1=CC=C(C(N2C(C3=CC=CC=C3)CN(C4=CC=C(C)C=C4)CC2)=O)C=C1)=O
Molecular Weight (g/mol) 413.51
Formula Weight 413.51
Percent Purity 99.89%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.