Learn More
MK 677, TRC
Product Code. 30011931
Change view
Click to view available options
Quantity:
5 mg
25 mg
250 mg
Unit Size:
250mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011931 25 mg 25mg
30134073 250 mg 250mg
30008884 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011931 Supplier Toronto Research Chemicals Supplier No. TRCM42499025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 159752-10-0
Synonym L-163,191∣MK-0677
Specifications
Purity >95% (HPLC)
Format Neat
Molecular Formula C28H40N4O8S2
MDL Number MFCD00942284
Molecular Weight (g/mol) 624.8, 624.77
InChI Key DUGMCDWNXXFHDE-VZYDHVRKSA-N
SMILES CS(O)(=O)=O.CC(C)(N)C(=O)N[C@H](COCC1=CC=CC=C1)C(=O)N1CCC2(CN(C3=CC=CC=C23)S(C)(=O)=O)CC1
IUPAC Name 2-amino-N-[(2R)-3-(benzyloxy)-1-{1-methanesulfonyl-1,2-dihydrospiro[indole-3,4'-piperidin]-1'-yl}-1-oxopropan-2-yl]-2-methylpropanamide; methanesulfonic acid
Product Type Impurity
Quantity 25 mg
Shipping Condition Room Temperature