missing translation for 'onlineSavingsMsg'
Learn More

Eprobemide, MedChemExpress

Product Code. 16423806
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16423806 25 mg 25mg
16498698 10 mg 10mg
16481108 5 mg 5mg
16444597 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16423806 Supplier Medchem Express Supplier No. HYB1413

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Eprobemide is a non-competitive reversible inhibitor of monoamine oxidase A.

Eprobemide is a non-competitive reversible inhibitor of monoamine oxidase A.

Specifications

Chemical Name or Material Eprobemide
CAS 87940-60-1
Color White
Purity Grade Notes Research
Molecular Formula C14H19ClN2O2
Quantity 25 mg
Synonym LIS 630
Solubility Information DMSO : ≥ 125 mg/mL (442.06 mM)
SMILES O=C(NCCCN1CCOCC1)C2=CC=C(Cl)C=C2
Molecular Weight (g/mol) 282.77
Formula Weight 282.77
Percent Purity 99.63%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.