missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calf thymus DNA, MedChemExpress
Description
Calf thymus DNA (DNA from calf thymus) is high quality double-stranded template DNA isolated from the thymus of male and female calves.
Specifications
Specifications
| Chemical Name or Material | Calf thymus DNA |
| Physical Form | Powder |
| CAS | 91080-16-9 |
| Color | White |
| Quantity | 25 mg |
| Purity Grade Notes | Research |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | DNA from calf thymus, Thymonucleic acid |
| Solubility Information | H2O : 20 mg/mL (Need ultrasonic) |
| SMILES | [Calf thymus DNA] |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?