missing translation for 'onlineSavingsMsg'
Learn More

AZD5153 6-Hydroxy-2-naphthoic acid, MedChemExpress

Product Code. 16472626
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16472626 25 mg 25mg
16479666 50 mg 50mg
16455847 5 mg 5mg
16413118 10 mg 10mg
16464406 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16472626 Supplier Medchem Express Supplier No. HY100653A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZD5153 6-Hydroxy-2-naphthoic acid is the 6-Hydroxy-2-naphthoic acid of AZD5153. AZD5153 is a potent, selective, and orally available BET/BRD4 bromodomain inhibitor; disrupts BRD4 with an IC50 of 1.7 nM.

AZD5153 6-Hydroxy-2-naphthoic acid is the 6-Hydroxy-2-naphthoic acid of AZD5153. AZD5153 is a potent, selective, and orally available BET/BRD4 bromodomain inhibitor

Specifications

Chemical Name or Material AZD5153 6-Hydroxy-2-naphthoic acid
CAS 1869912-40-2
Color White
Purity Grade Notes Research
Molecular Formula C36H41N7O6
Quantity 25 mg
Synonym AZD-5153 HNT salt
Solubility Information DMSO : ≥ 34 mg/mL (50.92 mM)
SMILES O=C(C1=CC=C2C=C(O)C=CC2=C1)O.O=C3N(C)CCN(CCOC4=CC=C(C5CCN(C6=NN7C(C=C6)=NN=C7OC)CC5)C=C4)[C@@H]3C
Molecular Weight (g/mol) 667.75
Formula Weight 667.75
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.