Learn More
Fisher Chemical 25KG BORIC ACID 99.8 % FOR ANALYSIS
Brand:  Fisher Chemical B/3800/70
25KG BORIC ACID 99.8 % FOR ANALYSIS
Product Code. 10325600
  • 5715.00 NOK / 25kg
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Boric acid
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.