missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone Isocaproate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Testosterone Isocaproate
Specifications
Specifications
| Chemical Name or Material | Testosterone isocaproate |
| CAS | 15262-86-9 |
| Analyte or Component Names | Testosterone isocaproate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H38 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Testosterone, 4-methylvalerate (6CI,8CI), Valeric acid, 4-methyl-, ester with testosterone (8CI), NSC 26641, Testosterone 17-isocaproate, Testosterone 4-methylpentanoate, Testosterone isocaproate, Androst-4-en-3-one, 17-[(4-methyl-1-oxopentyl)oxy]-, (17beta)-, (17beta)-17-[(4-Methyl-1-oxopentyl)oxy]androst-4-en-3-one, 3-Oxoandrost-4-en-17beta-yl isocaproate |
| InChI Formula | InChI=1S/C25H38O3/c1-16(2)5-10-23(27)28-22-9-8-20-19-7-6-17-15-18(26)11-13-24(17,3)21(19)12-14-25(20,22)4/h15-16,19-22H,5-14H2,1-4H3/t19-,20-,21-,22-,24-,25-/m0/s1 |
| SMILES | CC(C)CCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] 4-methylpentanoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction