missing translation for 'onlineSavingsMsg'
Learn More

Pyronaridine tetraphosphate, MedChemExpress

Product Code. 16499855
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499855 250 mg 250mg
16482287 50 mg 50mg
16481746 500 mg 500mg
16476536 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499855 Supplier Medchem Express Supplier No. HY14749A
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pyronaridine tetraphosphate is a Mannich base anti-malarial with demonstrated efficacy against drug resistant Plasmodium falciparum, P. vivax, P. ovale and P. malariae.

Pyronaridine tetraphosphate is a Mannich base anti-malarial with demonstrated efficacy against drug resistant Plasmodium falciparum, P. vivax, P. ovale and P. malariae.

Specifications

Chemical Name or Material Pyronaridine tetraphosphate
Physical Form Solid
CAS 76748-86-2
Color Yellow
Quantity 250 mg
Purity Grade Notes Research
Molecular Formula C29H44ClN5O18P4
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : 25 mg/mL (27.47 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 12.5 mg/mL (13.74 mM; ultrasonic and warming and heat to 60°C)
SMILES OC1=C(CN2CCCC2)C=C(NC3=C4N=C(OC)C=CC4=NC5=CC(Cl)=CC=C53)C=C1CN6CCCC6.O=P(O)(O)O.O=P(O)(O)O.O=P(O)(O)O.O=P(O)(O)O
Molecular Weight (g/mol) 910.03
Formula Weight 910.03
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.