Learn More
Wheaton™ 250 mL Polystyrene Sample Container
Sterile sample container offers sample security and user safety with leak free performance.
Brand: Wheaton™ 360252
697.54 NOK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 2.64000kg
Description
- For general sample collection, transport, and storage
- Wide container range to suit most user needs
Specifications
Specimen Container | |
Polystyrene | |
250 mL | |
Metal | |
63.4 mm | |
Plain | |
25/Pk. |
Yes | |
Sterile | |
EU Directive 98/79/EC | |
Screw Cap | |
121.2 mm | |
50/Cs. |