missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies 245 mm x 245 mm Square Treated Tissue Culture Dishes

Product Code. 17158061
Encompass_Preferred
Change view
Click to view available options
Quantity:
16 Dishes/Cs.
Unit Size:
Pack of 16
Product Code. Quantity unitSize
17158061 16 Dishes/Cs. Pack of 16
1 options
This item is not returnable. View return policy

Product Code. 17158061

Brand: STEMCELL Technologies 27141

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Intended for culturing anchorage-dependent cells

  • Tissue culture dishes, 245 mm x 245 mm, intended for culturing anchorage-dependent cells

Specifications

For Use With (Application) Cell Culture
Quantity 16 Dishes/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.