missing translation for 'onlineSavingsMsg'
Learn More

Spectrometric Oil Standard D12-010: Ag, Al, Cr, Cu, Fe, Mg, Na, Ni, Pb, Si, Sn, Ti @ 10 μg/g in Aviation Reference Oil, VHG™

Product Code. 30040804
Click to view available options
Quantity:
200 g
Unit Size:
240mL
This item is not returnable. View return policy

Product Code. 30040804

Brand: VHG Labs VHG9150001795145

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Spectrometric Oil Standard D12-010: Ag, Al, Cr, Cu, Fe, Mg, Na, Ni, Pb, Si, Sn, Ti @ 10 μg/g in Aviation Reference Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminium/Chromium/Copper/Iron/Lead/Magnesium/Nickel/Silicon/Silver/Sodium/Tin/Titanium
CAS 7429-90-5, 7440-47-3, 7440-50-8, 7439-89-6, 7439-92-1, 7439-95-4
Methods RDE
Analyte or Component Names Aluminium/Chromium/Copper/Iron/Lead/Magnesium/Nickel/Silicon/Silver/Sodium/Tin/Titanium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Aviation Oil
Quantity 200 g
Concentration or Composition (by Analyte or Components) Aluminium 10 μg/g/Chromium 10 μg/g/Copper 10 μg/g/Iron 10 μg/g/Lead 10 μg/g/Magnesium 10 μg/g/Nickel 10 μg/g/Silicon 10 μg/g/Silver 10 μg/g/Sodium 10 μg/g/Tin 10 μg/g/Titanium 10 μg/g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.