Learn More
Macherey-Nagel™ Weighing Papers
Product Code. 10222231 Shop All Macherey-Nagel Products
Change view
Click to view available options
Dimensions (L x W):
90 x 115 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
10222231 90 x 115 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10222231 Supplier Macherey-Nagel™ Supplier No. 186002
Only null left
Add to basket
Add to basket
Guarantees that weighed goods can be transferred without loss. Macherey Nagel™ Weighing Papers are transparent and smooth on both sides.
  • Grade MN 226
  • Can be used as a substitute for weighing boats
Specifications
Dimensions (L x W) 90 x 115 mm
Length (Metric) 90 mm
Quantity 100
Width (Metric) 115 mm
Product Type Weighing Paper
Shape Boat