Learn More
KGW Isotherm™ 21000mL Large Insulating Dewar Flasks
Click to view available options
Capacity (Metric):
21 L
Unit Size:
Each
Specifications
Specifications
Diameter (Metric) Outer | 280mm |
For Use With (Application) | Storage and transport of temperature sensitive goods e.g. dry ice ( CO2 ) |
Height (Metric) Inner | 480mm |
Height (Metric) | 615mm |
Includes | Quick release clamp |
Capacity (Metric) | 21 L |
Diameter (Metric) Inner | 250 mm |
Material | Borosilicate Glass |
Product Content Correction
Product Title
KGW Isotherm™ 21000mL Large Insulating Dewar Flasks >