Learn More
Invitrogen™ 20X Bolt™ MES SDS Running Buffer
Product Code. 13266499
Change view
Click to view available options
Quantity:
500 mL
5 L
Unit Size:
5 L
500mL
Product Code. Quantity unitSize
13266499 500 mL 500mL
13246499 5 L 5 L
2 options
Product Code. 13266499
Brand: Invitrogen™ B0002
Optimized for use with Bolt™ Bis-Tris Plus gels, and are available in a variety of formats
About Bolt™ Bis-Tris Plus Gels
Bolt™ Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of your small- to medium-sized proteins under denaturing conditions. Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt™ gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt™ gels to obtain optimal results for your day-to-day protein separation needs. Bolt™ Bis-Tris Plus gels come in four gel types and multiple well formats. A Mini Gel Tank is required for use of all Bolt™ gels.
Bolt™ gel run times may vary depending upon the type of running buffer used.
Order Info
Shelf-life is 6 months, when stored properly.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material MES SDS Running Buffer
Concentration 20X
For Use With (Equipment) Bolt
Product Line Bolt
Quantity 500 mL