Description
Quick and easy assembly and disassembly without the need for tools. Barbed ridges provide secure grip on the inside of fle in., ible tubing. Constructed of USP compliant polypropylene
Specifications
Specifications
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Packaging Quantity | 25/Pk. |
| Outlet Port | Hose Barb Y |
| Sterilization Method | Gamma radiation, ethylene oxide (EtO), E-beam, and autoclave |
| Shape | Y |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.