Learn More
Abnova™ 2 x HotTaq PCR Mix with MgCl2
Product Code. 16022565
Change view
Click to view available options
Quantity:
1 Tub
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16022565 1 Tub Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16022565 Supplier Abnova™ Supplier No. D0017.1Tub
 Restricted Item
Estimated Shipment: 05-02-2027
HotTaq PCR mix is a prepared solution containing everything needed for successful PCR reaction except specific primers and DNA template.
  • The mix includes highquality recombinant Taq DNA polymerase, nucleotides in a specifically optimized buffer formulation.
  • For reaction set-up add the PCR Mix (25 ul) to the primers, template and water (is provided in the set; total volume of 50 ul).
  • PCR Mix is provided with MgCl2 in the solution.
Specifications
Content And Storage Store at -20°C Guaranteed stable for 6 months when properly stored
Quantity 1 Tub
For Use With (Application) Genotyping