Learn More
Ohaus™ 2 x Centrifuge Tube Adapters, Frontier™ R-A6x250/8MI Fixed Rotors
Shop All Ohaus Products
Click to view available options
Capacity:
1 x 50 mL (29 mm dia.)
1 x 50 mL (30 mm dia)
2 x 30 mL
4 x 15 mL
5 x 10 mL
8 x 1.5 mL
Unit Size:
Pack of 2
Description
- Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
- Suitable for use in Frontier™ R-A6x250/8MI Fixed Rotors
Specifications
Specifications
| Type | RB |
| Capacity | 1 x 50 mL (29 mm dia.) |
| Max. Tube Diameter | 29 mm |
| For Use With (Equipment) | R-A6x250/8MI Fixed Rotors |
| Description | Adapter, 1x50ml D29mm RB, 2/pk |
| No. of Places | 1 |
| No. per Pack | 2 |