Learn More
Ohaus™ 2 x Centrifuge Tube Adapters, Frontier™ Fixed Angle Rotors
Product Code. 15806732 Shop All Ohaus Products
Click to view available options
Capacity:
1 x 15 mL (17 mm dia. FA)
1 x 15 mL (17 mm dia. RB)
1 x 16 mL
1 x 30 mL
1 x 50 mL (29 mm dia. FA)
1 x 50 mL (29 mm dia. RB)
2 x 15 mL
Unit Size:
Pack of 2
Product Code. 15806732
Brand: Ohaus™ 30314894
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ Fixed Angle Rotors.
  • Accommodates centrifuge tubes that are smaller and/or greater in number than the space provided in the rotor carrier
  • Suitable for use in the following Frontier™ rotors:
    • R-A4x85/12MI
    • R-A6x85/13MI
    • R-A6x85/14MI
TRUSTED_SUSTAINABILITY
Specifications
Type RB
Capacity 1 x 30 mL
Max. Tube Diameter 25 mm
For Use With (Equipment) Frontier Fixed-Angle Rotors
Description Adapter, 1x30ml D25mm RB, 2/pk
No. of Places 1
No. per Pack 2