Description
- Allows you to centrifuge tubes of custom size or number
- Suitable for use in buckets for Frontier™ R-S4x100/5MIS Swing Out Rotors
Specifications
Specifications
| Capacity | 1 x 50 mL (29 mm dia. Rim) |
| Max. Tube Diameter | 29 |
| For Use With (Equipment) | R-S4x100/5MIS Swing Out Rotors |
| Description | Rack, 1x50ml D29mm Rim, 2/pk, V2 |
| No. of Places | 1 |
| No. per Pack | 2 |