Learn More
Ohaus™ 2 x Centrifuge Rack Adapters, Frontier™ R-S4x100/4MI Swing Out Rotors
Product Code. 15816692 Shop All Ohaus Products
Click to view available options
Capacity:
1 x 100 mL
1 x 15 mL
1 x 25 mL
1 x 30 mL
1 x 50 mL (29 mm dia.)
1 x 50 mL (29 mm dia., FA)
1 x 50 mL (29 mm dia.,RB)
1 x 50 mL (34 mm dia.)
1 x 85 mL
10 x 1.5 mL
2 x 15 mL
5 x 4 to 10 mL
7 x 15 mL
7 x 5 to 7 mL (13 mm dia., RB)
7 x 5 to 7 mL (13 mm dia., Vac)
9 x 4.5 mL
9 x 5 to 7 mL
Unit Size:
Pack of 2
Product Code. 15816692
Brand: Ohaus™ 30314854
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ R-S4x100/4MI Swing Out Rotors
  • Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
  • Suitable for use in Frontier™ R-S4x100/4MI Swing Out Rotors
TRUSTED_SUSTAINABILITY
Specifications
Capacity 10 x 1.5 mL
Max. Tube Diameter 11 mm
For Use With (Equipment) R-S4x100/4MI Swing Out Rotors
Description Rack, 10x1.5ml D11mm, 2/pk
No. of Places 10
No. per Pack 2