Learn More
Ohaus™ 2 x Centrifuge Rack Adapters, Frontier™ R-S16x50/4MI Swing Out Rotors
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ R-S16x50/4MI Swing Out Rotors
Brand:  Ohaus™ 30314583
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15856652
  • 3355.00 NOK / Pack of 2
In Stock
Description
  • Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
  • Suitable for use in Frontier™ R-S16x50/4MI Swing Out Rotors
Specifications
Specifications
FA
17 mm
Rack, 10x15ml D17mm FA, 2/pk
2
10 x 15 mL
R-S16x50/4MI Swing Out Rotors
10