Learn More
Ohaus™ 2 x Centrifuge Buckets, Frontier™ R-S4x100/5MIS Swing Out Rotor
Product Code. p-7158845 Shop All Ohaus Products
Click to view available options
Capacity:
1 x 100 mL (w/ cap)
1 x 100 mL (w/o cap)
2 x 50 mL
3 x 15 mL
Unit Size:
Pack of 2
Product Code. 15866692
Brand: Ohaus™ 30314860
Carrier buckets for centrifuge tubes, suitable for the R-S4x100/5MIS Swing Out Rotor.
  • Swinging bucket holds specifically sized tubes for centrifugation
  • Suitable for use in Frontier™ R-S4x100/5MIS Swing Out Rotors
TRUSTED_SUSTAINABILITY
Specifications
Description Bucket, 100ml w/o Cap, 2/pk
Capacity 1 x 100 mL (w/ cap)
For Use With (Equipment) R-S4x100/5MIS Swing Out Rotor
Includes 2 Buckets
No. of Places 1