Learn More
2-(Trimethylsiloxy)-1,3-butadiene, Sigma-Aldrich™
Specifications
Specifications
| CAS | 38053-91-7 |
| Density | 0.82 g/cm3 at 20°C |
| Boiling Point | 124°C/1013 hPa |
| Molecular Formula | (CH3)3SiOC4H5 |
| MDL Number | MFCD00009852 |
| Quantity | 10 mL |
| Synonym | 2-(Trimethylsiloxy)-1,3-butadiene, 2-(Trimethylsiloxy)-1,3-butadiene |
| Molecular Weight (g/mol) | 142.27 |
| Grade | Research |