Få mere at vide
2-(Trifluoromethyl)pyridine-5-boronic acid, 98%
Artikelnummer. p-7055506
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
250 mg
1 g
5 g
Pakningsstørrelse:
1g
250mg
5g
Artikelnummer. Quantity unitSize
15438706 250 mg 250mg
15431159 1 g 1g
15441159 5 g 5g
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15438706
Brand: Thermo Scientific Alfa Aesar H64106.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Denne vare kan ikke returneres. Se returpolicy
CAS: 868662-36-6 | C6H5BF3NO2 | 190.916 g/mol
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 868662-36-6
Molecular Formula C6H5BF3NO2
Molecular Weight (g/mol) 190.916
MDL Number MFCD07375381
InChI Key BNTIPMNMTIAWIW-UHFFFAOYSA-N
Synonym 2-trifluoromethyl pyridine-5-boronic acid, 6-trifluoromethyl pyridin-3-yl boronic acid, 2-trifluoromethyl-5-pyridineboric acid, 2-trifluoromethyl pyridin-5-ylboronic acid, 6-trifluoromethyl pyridine-3-boronic acid, 6-trifluoromethyl pyridin-3-ylboronic acid, 2-trifluoromethylpyridine-5-boronic acid, 2-trifluoromethyl-5-pyridineboronic acid, 6-trifluoromethyl-3-pyridylboronic acid
PubChem CID 25134278
IUPAC Name [6-(trifluoromethyl)pyridin-3-yl]boronic acid
SMILES B(C1=CN=C(C=C1)C(F)(F)F)(O)O
Specifica
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 190.92
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-(Trifluoromethyl)pyridine-5-boronic acid
RUO – Research Use Only
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.