Learn More
2-(Tri-n-butylstannyl)pyridine, tech. 80%
Product Code. p-7027860
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11481387
Brand: Thermo Scientific Alfa Aesar B24911.03
Request Bulk or Custom Format
CAS: 17997-47-6 | C17H31NSn | 368.152 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 17997-47-6
Molecular Formula C17H31NSn
Molecular Weight (g/mol) 368.152
MDL Number MFCD00052052
InChI Key GYUURHMITDQTRU-UHFFFAOYSA-N
Synonym 2-tributylstannyl pyridine, 2-tri-n-butylstannylpyridine, 2-tributylstannylpyridine, tributyl 2-pyridyl tin, 2-pyridyltributyltin, 2-1,1,1-tributylstannyl pyridine, 2-pyridyltri-n-butyltin, 2-tributylstannyl-pyridine, tri-n-butyl 2-pyridyl tin
PubChem CID 607784
IUPAC Name tributyl(pyridin-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CC=N1
Specifications
Density 1.15
Boiling Point 130°C to 132°C (0.8 mmHg)
Refractive Index 1.513
Quantity 1 g
UN Number UN2788
Beilstein 479329
Formula Weight 368.15
Percent Purity 80%
Grade Technical
Chemical Name or Material 2-(Tri-n-butylstannyl)pyridine