Learn More
2-(Tri-n-butylstannyl)oxazole, 95%
Product Code. 15412258
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 15412258
Brand: Thermo Scientific Alfa Aesar H55870.03
Request Bulk or Custom Format
CAS: 145214-05-7 | C15H29NOSn | 358.113 g/mol
2-(Tri-n-butylstannyl)oxazole act as a synthetic building block, which is utilized in Stille coupling
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Tri-n-butylstannyl)oxazole act as a synthetic building block, which is utilized in Stille coupling
Solubility
Insoluble in water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 145214-05-7
Molecular Formula C15H29NOSn
Molecular Weight (g/mol) 358.113
MDL Number MFCD06798076
InChI Key YOWGRWHKDCHINP-UHFFFAOYSA-N
Synonym 2-tributylstannyl oxazole, 2-tri-n-butylstannyl oxazole, 2-tributylstannyl-1,3-oxazole, 2-tributylstannanyl-oxazole, 2-tributylstannyloxazole, 2tributylstannyloxazole, acmc-20aom1, 2-tributylstannyl-oxazole, 2-tributylstannanyloxazole
PubChem CID 15486183
IUPAC Name tributyl(1,3-oxazol-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=NC=CO1
Specifications
Color Colorless
Density 1.71 g/mL
Boiling Point 108°C to 110°C (0.2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.493
Quantity 1 g
UN Number UN2788
Solubility Information Insoluble in water.
Formula Weight 358.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-(Tri-n-butylstannyl)oxazole