Description
- Whether in the assembly hall or on the production line, indoors or outdoors.
- uvex 2 trend safety footwear range is suitable for medium-risk applications (S1-S3) and incorporates numerous proven technological safety and comfort features at an affordable price.
- The uvex anti-twist heel cap offers increased stability without limiting movement to protect against twists and sprains.
- The high quality outer sole materials, sports influence look and bold colors make this footwear appealing to wear.
Specifications
Specifications
| Color | Black, White, Yellow |
| Material | Polyurethane |
| Size | 35 |
| Disposable | Reusable |
| Gender | Unisex |
| Waterproof | Water Resistant |
| Slip Resistance Rating | SRC |
| Product Type | Safety Shoes |