Learn More
2-Thiopheneacetic Acid, TRC
Product Code. 30454423
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454423 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30454423 Supplier Toronto Research Chemicals Supplier No. TRCT34479550G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Thiopheneacetic Acid
CAS 1918-77-0
Molecular Formula C6H6O2S
Quantity 50 g
Synonym 2-(2-Thienyl)acetic Acid, 2-(Thien-2-yl)acetic Acid, 2-(Thiophen-2-yl)acetic Acid, 2-Thienylacetic Acid, 2-Thienylethanoic Acid, Thiophen-2-ylacetic Acid
InChI Formula InChI=1S/C6H6O2S/c7-6(8)4-5-2-1-3-9-5/h1-3H,4H2,(H,7,8)
SMILES O=C(O)CC1=CC=CS1
IUPAC Name 2-thiophen-2-ylacetic acid
Molecular Weight (g/mol) 142.18
Formula Weight 142.0089