Learn More
2-(tert-Butylamino)propiophenone Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
2-(tert-Butylamino)propiophenone Hydrochloride
Specifications
Specifications
| Chemical Name or Material | 2-(tert-Butylamino)propiophenone Hydrochloride |
| CAS | 63199-74-6 |
| Analyte or Component Names | 2-(tert-Butylamino)propiophenone Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H19 N O . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-(tert-Butylamino)propiophenone Hydrochloride, 282U64 hydrochloride, SKF-44633-A, Bupropion related, 2-(t-butylamino)propiophenone HCl (282U64 HCl) |
| InChI Formula | InChI=1S/C13H19NO.ClH/c1-10(14-13(2,3)4)12(15)11-8-6-5-7-9-11;/h5-10,14H,1-4H3;1H |
| SMILES | Cl.CC(NC(C)(C)C)C(=O)c1ccccc1 |
| IUPAC Name | 2-(tert-butylamino)-1-phenylpropan-1-one;hydrochloride |
| Show More |