Learn More
2-Sulfobenzoic Acid Ammonium Salt, Sigma-Aldrich™
Specifications
Specifications
| CAS | 6939-89-5 |
| Melting Point | 274°C to 277°C |
| Molecular Formula | 2-(HOOC)C6H4SO3NH4 |
| MDL Number | MFCD00040590 |
| Quantity | 25 g |
| Synonym | 2-Sulfobenzoic acid ammonium salt, 2-Sulfobenzoic acid ammonium salt |
| Molecular Weight (g/mol) | 219.22 |
| Grade | Research |