Learn More
2-sec-Butyl-d5-4-{4-[4-(4-methyloxy-phenyl)-piperazin-1-yl]-phenyl}-2,4-dihydro-[1,2,4]-triazol-3-one, TRC
Specifications
Specifications
| Chemical Name or Material | 2-sec-Butyl-d5-4-{4-[4-(4-methyloxy-phenyl)-piperazin-1-yl]- |
| CAS | 1020719-22-5 |
| Molecular Formula | C23 2H5 H24 N5 O2 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C23H29N5O2/c1-4-18(2)28-23(29)27(17-24-28)21-7-5-19(6-8-21)25-13-15-26(16-14-25)20-9-11-22(30-3)12-10-20/h5-12,17-18H,4,13-16H2,1-3H3/i1D3,4D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])C(C)N1N=CN(C1=O)c2ccc(cc2)N3CCN(CC3)c4ccc(OC)cc4 |
| IUPAC Name | 4-[4-[4-(4-methoxyphenyl)piperazin-1-yl]phenyl]-2-(3,3,4,4,4-pentadeuteriobutan-2-yl)-1,2,4-triazol-3-one |
| Molecular Weight (g/mol) | 412.54 |
| Formula Weight | 412.2635 |