missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, VLSI Grade, Scharlab

Product Code. 11352445
Change view
Click to view available options
Quantity:
25 L
Packaging:
Stainless Steel Drum
Unit Size:
25 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11352445 25 L Stainless Steel Drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11352445 Supplier Scharlab Supplier No. AL0322025B

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

2-Propanol VLSI Grade, Scharlab is guaranteed to be free of water, metals and particles.

VLSI (Very Large Scale Integration) Grade solvents are often used in the semiconductor industry and in electronic industry production, mainly in the cleaning of integrated circuits.

Specifications

Melting Point -89.5°C
CAS 67-63-0
Color Colorless
Acidity Max. 0.0002meq/g
Alkalinity Max. 0.0001meq/g
Density 0.78g/cm3
Quantity 25 L
Flash Point 12°C
Identification Passes test
Ignition Point 425°C
Packaging Stainless Steel Drum
Physical Form Liquid
Residue after Evaporation Max. 0.0005%
Molecular Formula C3H8O
Vapor Pressure 43 hPa
UN Number 1219
Water Max. 0.05%
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Boiling Point 82.4°C
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES CC(C)O
IUPAC Name propan-2-ol
Molecular Weight (g/mol) 60.096
PubChem CID 3776
ChEBI CHEBI:17824
Formula Weight 60.10g/mol
Percent Purity Min. 99.8%
Grade LCSI
Chemical Name or Material 2-Propanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.