missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, HPLC Gradient Grade, Scharlab

Catalog Number 11372445
Click to view available options
Quantity:
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Suitable for use in solvents for liquid chromatography, in antifreeze compositions and in cosmetics.

Microfiltered through membranes of pore diameter 0.22μm and suitable for UPLC.

  • Synonyms: Isopropyl alcohol, Isopropanol, iso-Propanol, Dimethylcarbinol, 2-Hydroxypropane
  • Molecular Formula: C3H8O
  • Molar Mass: 60.10g/mol
  • CAS: 67-63-0
  • EIN ECS No.: 200-661-7
  • Dielectric Constant: 18.3 at 25°C
Melting Point -89.5°C
Absorbance Max background: 0.025AU at 240nm
CAS 67-63-0
Color Colorless
Acidity 0.0001mEq/g max
Alkalinity 0.0001mEq/g max
Density 0.78g/cm3
Boiling Point 82.4°C
Quantity 2.5 L
Flash Point 12°C
Identification Passes Test
Ignition Point 425°C
Physical Form Liquid
Residue after Evaporation 0.0001% max
Molecular Formula C3H8O
Vapor Pressure 43hPa at 20°C
Water 0.05% max
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES CC(C)O
IUPAC Name propan-2-ol
Molecular Weight (g/mol) 60.096
PubChem CID 3776
ChEBI CHEBI:17824
Formula Weight 60.1g/mol
Percent Purity 99.9% min
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Propanol, gradient HPLC grade
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'