Learn More
Cole-Parmer™ Essentials Water Bath Clamp, Two-Prong, Small
Product Code. 11725469 Shop All Cole-Parmer Products
Change view
Click to view available options
Size (Metric) Grip:
4.57 to 9.91 mm
Unit Size:
Each
Product Code. Size (Metric) Grip unitSize
11725469 4.57 to 9.91 mm Each
1 options
Product Code. 11725469
Brand: Cole-Parmer™ Essentials 0804312
Only null left
Add to basket
Add to basket
Provide support for your specific laboratory equipment Small, Two-Prong, Water Bath Clamp.
  • These C-shaped clamps with thumbscrew attach securely to your bath wall.
  • 0.39 in. Max Grip, 0.43 in. Max Tank Wall.
Specifications
Quantity 1 Ea.
Type Water Bath Clamp
Material Nickel-plated Zinc
For Use With (Equipment) Water Bath
Diameter (English) 0.2 in. to 0.4 in.
Diameter (Metric) 5 to 10 mm
Length (English) 3.5 in.
Length (Metric) 88.9 mm
Size Small
Size (Metric) Grip 4.57 to 9.91 mm
Size (English) Grip 0.18 to 0.39 in.
Shape C-shaped
Jaw Type 2-Prong