missing translation for 'onlineSavingsMsg'
Learn More

2-Picolinic Acid HCl, MP Biomedicals

Product Code. 11453595 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453595 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11453595 Supplier MP Biomedicals Supplier No. 0521682010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 636-79-3
Molecular Formula C6H6ClNO2
Quantity 10 g
Synonym niacin hydrochloride, nicotinic acid hydrochloride, 3-pyridinecarboxylic acid, hydrochloride, nicotinic acid, hydrochloride, pyridine-3-carboxylic acid hydrochloride, 3-pyridinecarboxylic acid, hydrochloride 1:1, 59-67-6 parent, niacin hcl, nicotinic acid hydrochloride 1:1, 3-pyridinecarboxylic acid hydrochloride
InChI Key IBTWUVRCFHJPKN-UHFFFAOYSA-N
SMILES C1=CC(=CN=C1)C(=O)O.Cl
IUPAC Name pyridine-3-carboxylic acid;hydrochloride
Molecular Weight (g/mol) 159.569
PubChem CID 71558
Chemical Name or Material 2-Picolinic Acid HCL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.