Learn More
2-Phenyl-1,3-thiazole-4-carbonyl chloride, Thermo Scientific™
Product Code. 10019191
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10019191 250 mg 250mg
10121112 1 g 1g
10103344 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10019191 Supplier Thermo Scientific Maybridge Supplier No. CC18302CB
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Chemical Identifiers
CAS 36094-04-9
Molecular Formula C10H6ClNOS
Molecular Weight (g/mol) 223.674
InChI Key ZZFOIDMEHXGBKS-UHFFFAOYSA-N
Synonym 2-phenylthiazole-4-carbonyl chloride, 2-phenyl-thiazole-4-carbonyl chloride, 4-thiazolecarbonylchloride, 2-phenyl
PubChem CID 2776472
IUPAC Name 2-phenyl-1,3-thiazole-4-carbonyl chloride
SMILES C1=CC=C(C=C1)C2=NC(=CS2)C(=O)Cl
Specifications
Assay Percent Range 95%
Quantity 250 mg
Formula Weight 223.68
Percent Purity 95%
Chemical Name or Material 2-Phenyl-1, 3-thiazole-4-carbonyl chloride