missing translation for 'onlineSavingsMsg'
Learn More

2-Pentanol, Sigma-Aldrich™

Product Code. 15866460
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15866460 100 mL 100mL
15829500 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15866460 Supplier Sigma-Aldrich Supplier No. 8075010100
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6032-29-7
Grade Research
Molecular Formula CH3CH2CH2CH(OH)CH3
Molecular Weight (g/mol) 88.15

Specifications

Melting Point -50°C
Density 0.81 g/cm3 at 20°C
Boiling Point 119°C/1013 hPa
MDL Number MFCD00004579
Quantity 100 mL
Synonym 2-Pentanol, Methyl propyl carbinol, sec-n-Amyl alcohol, 2-Pentanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.