missing translation for 'onlineSavingsMsg'
Learn More

(2-Oxopropyl)carbamic Acid tert-Butyl Ester, TRC

Product Code. 30428567
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Product Code. Quantity
30428567 100 mg
30414346 500 mg
30424578 50 mg
3 options
This item is not returnable. View return policy

Product Code. 30428567

Brand: Toronto Research Chemicals TRCO990068100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170384-29-9
Formula Weight 173.105
InChI Formula InChI=1S/C8H15NO3/c1-6(10)5-9-7(11)12-8(2,3)4/h5H2,1-4H3,(H,9,11)
IUPAC Name tert-butyl N-(2-oxopropyl)carbamate
Molecular Formula C8 H15 N O3
Molecular Weight (g/mol) 173.21
SMILES CC(=O)CNC(=O)OC(C)(C)C
Chemical Name or Material (2-Oxopropyl)carbamic Acid tert-Butyl Ester

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.