Learn More
2-Oxohexanoic Acid, TRC
Product Code. 30423205
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30423205 500 mg
30409857 1 g
30415034 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30423205 Supplier Toronto Research Chemicals Supplier No. TRCO991128500MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2492-75-3
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
Synonym 2-Ketocaproic Acid, 2-Ketohexanoic Acid, 2-Oxocaproic Acid, alpha-Ketocaproic Acid, alpha-Ketohexanoic Acid, alpha-Oxo-n-caproic Acid, alpha-Oxocaproic Acid
IUPAC Name 2-oxohexanoic acid
SMILES CCCCC(=O)C(=O)O
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C6H10O3/c1-2-3-4-5(7)6(8)9/h2-4H2,1H3,(H,8,9)
Formula Weight 130.063
Chemical Name or Material 2-Oxohexanoic Acid