Learn More
2-Oxa-6-azaspiro[3.3]heptane oxalate, 97%
Product Code. 15460339
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15460339 5 g 5g
15470339 250 mg 250mg
15450339 1 g 1g
3 options
Product Code. 15460339
Brand: Thermo Scientific Alfa Aesar H63578.06
Request Bulk or Custom Format
CAS: 1159599-99-1 | C7H11NO5 | 189.17 g/mol
2-Oxa-6-azaspiro[3.3]heptane oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Oxa-6-azaspiro[3.3]heptane oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1159599-99-1
Molecular Formula C7H11NO5
Molecular Weight (g/mol) 189.17
MDL Number MFCD11976178
InChI Key KOUVDKDABFOPIG-UHFFFAOYSA-N
Synonym 2-oxa-6-azaspiro 3.3 heptane oxalate, 2-oxa-6-aza-spiro 3.3 heptane oxalic acid salt, 2-oxa-6-azaspiro 3,3 heptane oxalic acid salt, 2-oxa-6-azaspiro 3.3 heptane xoxalate, 2-oxa-6-azaspiro 3.3 heptane oxalic acid, 2-oxa-6-azaspiro 3.3 heptane; oxalic acid, 2-oxa-6-azaspiro 3.3 heptane ethanedioate, 2-oxa-6-azaspiro 3.3 heptane oxalate 1:1, 2-oxa-6-azaspiro 3.3 heptane, ethanedioate 1:1, acmc-2099rj
PubChem CID 53308464
SMILES OC(=O)C(O)=O.C1NCC11COC1
Specifications
Quantity 5 g
Solubility Information Slightly soluble in water.
IUPAC Name 2-oxa-6-azaspiro[3.3]heptane; oxalic acid
Formula Weight 189.17
Percent Purity 97%
Chemical Name or Material 2-Oxa-6-azaspiro[3.3]heptane oxalate