Learn More
2-Nonadecanone, tech. 80%
Product Code. 11424677
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11424677
Brand: Thermo Scientific Alfa Aesar B21178.14
Request Bulk or Custom Format
CAS: 629-66-3 | C19H38O | 282.51 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 629-66-3
Molecular Formula C19H38O
Molecular Weight (g/mol) 282.51
MDL Number MFCD00026671
InChI Key IEDKVDCIEARIIU-UHFFFAOYSA-N
Synonym 2-nonadecanone, nonadeca-2-one, methyl heptadecyl ketone, acmc-20anop, 2-oxononadecan-3-yl, n-heptadecyl methyl ketone
PubChem CID 69423
IUPAC Name nonadecan-2-one
SMILES CCCCCCCCCCCCCCCCCC(C)=O
Specifications
Melting Point 54°C to 56°C
Flash Point >100°C (212°F)
Linear Formula CH3(CH2)16COCH3
Quantity 25 g
Beilstein 1708703
Formula Weight 282.51
Percent Purity 80%
Grade Technical
Chemical Name or Material 2-Nonadecanone