Learn More
2-Nitrotoluene, Sigma-Aldrich™
Product Code. 15839340
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15839340 1 L 1 L
15829340 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15839340 Supplier Sigma Aldrich Supplier No. 8068061000
 Restricted Item
Estimated Shipment: 20-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 88-72-2
Grade Research
Molecular Formula 2-(NO2)C6H4CH3
Molecular Weight (g/mol) 137.14
Specifications
Melting Point -9°C
Density 1.16 g/cm3 at 20°C
Boiling Point 221°C to 222°C/1013 hPa
MDL Number MFCD00007157
Quantity 1 L
Synonym 2-Nitrotoluene, o-Nitrotoluene, 2-Nitrotoluene