missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenyl beta-D-glucopyranoside, 98+%, Thermo Scientific™

Product Code. 11412767
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412767 1 g 1g
11422767 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11412767 Supplier Thermo Scientific Alfa Aesar Supplier No. A14935.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Nitrophenyl beta-D-glucopyranoside is used as a substrate for ß -glucosidase.

Solubility
Soluble in water

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2816-24-2
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.25
MDL Number MFCD00065048,MFCD00063255,MFCD00065048,MFCD00065048
InChI Key KUWPCJHYPSUOFW-UHFFFAOYNA-N
Synonym 2-nitrophenyl b-d-glucopyranoside
PubChem CID 92211304
IUPAC Name (2R,3S,4R,5R,6S)-2-(hydroxymethyl)-6-(2-nitrophenoxy)oxane-3,4,5-triol
SMILES OCC1OC(OC2=CC=CC=C2[N+]([O-])=O)C(O)C(O)C1O

Specifications

Melting Point 141°C to 142°C
Quantity 1 g
Beilstein 92206
Solubility Information Soluble in water
Optical Rotation −105° (c=2 in Water)
Formula Weight 301.25
Percent Purity ≥98%
Chemical Name or Material 2-Nitrophenyl beta-D-glucopyranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.