Learn More
Melford 2-Nitrophenyl-B-D-glucopyranoside
Product Code. 30018619
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018619 10 g 10g
30018617 100 mg 100mg
30018618 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30018619 Supplier Melford Supplier No. N8102010.0
 Restricted Item
Estimated Shipment: 12-11-2026
Spectrophotometric substrate for detection of β-D-Glucosidase.
Spectrophotometric substrate for detection of β-D-Glucosidase.
Specifications
Quantity 10 g