missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 2-Nitrofurazone metabolite Monoclonal Antibody (5G2)
GREENER_CHOICE

Product Code. 17919422
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
Product Code. Quantity unitSize
17919422 200 μg 200µg
1 options
This item is not returnable. View return policy

Product Code. 17919422

Brand: Invitrogen™ MA549745

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

2-Nitrofurazone metabolite (2-NFZ) is a nitrofuran antibiotic used in veterinary medicine to treat a wide range of bacterial infections in animals. It is a metabolite of the drug nitrofurazone, which is commonly used in topical applications for wound care in animals. 2-NFZ works by inhibiting bacterial growth and is effective against a broad spectrum of gram-positive and gram-negative bacteria. It is generally well-tolerated by animals, with few side effects reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 2-Nitrofurazone metabolite
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 5G2
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of 2-CPSEM hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.