Learn More
2′-Nitroacetanilide, Sigma-Aldrich™
Specifications
Specifications
| CAS | 552-32-9 |
| Melting Point | 90°C to 93°C |
| MDL Number | MFCD00016991 |
| Quantity | 25 g |
| Synonym | 2′-Nitroacetanilide, 2′-Nitroacetanilide |
| Molecular Weight (g/mol) | 180.16 |
| Grade | Research |