Learn More
2-Nitro-5-(trifluoromethyl)aniline, TRC
Product Code. 30471172
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30471172 1 g
30482047 500 mg
30464759 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30471172 Supplier Toronto Research Chemicals Supplier No. TRCN5873901G
 Restricted Item
Estimated Shipment: 03-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 402-14-2
Molecular Formula C7 H5 F3 N2 O2
Molecular Weight (g/mol) 206.122
Synonym alpha,alpha,alpha-Trifluoro-6-nitro-m-toluidine, 2-Nitro-5-(trifluoromethyl)aniline, 3-Amino-4-nitrobenzotrifluoride, 5-Trifluoromethyl-2-nitroaniline, NSC 10324, 2-Nitro-5-(trifluoromethyl)-benzenamine
IUPAC Name 2-nitro-5-(trifluoromethyl)aniline
SMILES Nc1cc(ccc1[N+](=O)[O-])C(F)(F)F
Specifications
Quantity 1 g
InChI Formula InChI=1S/C7H5F3N2O2/c8-7(9,10)4-1-2-6(12(13)14)5(11)3-4/h1-3H,11H2
Formula Weight 206.03
Chemical Name or Material 2-Nitro-5-(trifluoromethyl)aniline