missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-5-(trifluoromethyl)aniline, 97%

Product Code. 11328735
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328735 5 g 5g
11318735 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328735 Supplier Thermo Scientific Alfa Aesar Supplier No. L12310.06
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 402-14-2 | C7H5F3N2O2 | 206.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-14-2
Molecular Formula C7H5F3N2O2
Molecular Weight (g/mol) 206.12
MDL Number MFCD00042447
InChI Key AUTLVHYEAAAKNM-UHFFFAOYSA-N
Synonym 2-nitro-5-trifluoromethyl aniline, 3-amino-4-nitrobenzotrifluoride, 3-amino-4-nitrobenzitrifluoride, benzenamine, 2-nitro-5-trifluoromethyl, 5-trifluoromethyl-2-nitrobenzenamine, 2-nitro-5-trifluoromethyl phenylamine, 2-nitro-5-trifluormethyl anilin, pubchem2790, 3-amino-4-nitrobenztrifluoride, 2-nitro-5-trifluoromethyl-aniline
PubChem CID 223100
IUPAC Name 2-nitro-5-(trifluoromethyl)aniline
SMILES NC1=CC(=CC=C1[N+]([O-])=O)C(F)(F)F

Specifications

Melting Point 98°C to 102°C
Quantity 5 g
UN Number UN2811
Beilstein 2732344
Formula Weight 206.13
Percent Purity 97%
Chemical Name or Material 2-Nitro-5-(trifluoromethyl)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.